Gene Transcripts

Gene Symbol
GPC6
Entrez Gene ID
10082
Gene Name
glypican 6
Gene Aliases
OMIMD1
Chromosome Location
Chr. 13: 93226825 - 94408020 on Build GRCh38
UniGene ID
Hs.444329
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005708.3 NP_005699.1 3 1148
XM_011521044.2 XP_011519346.1 3 473
XM_017020298.1 XP_016875787.1 3 2756
XM_017020299.1 XP_016875788.1 3 1523
XM_017020300.1 XP_016875789.1 3 380
XM_017020301.1 XP_016875790.1 2 351
GenBank mRNA AF105267.1 - 3 1101
AF111178.1 - 3 1150
AK074741.1 - 3 602
AK130013.1 - 3 485
AK290144.1 - 3 669
AK304338.1 - 3 967
AY358462.1 - 3 1081
BC106947.2 - 3 996 40034564

View Full Product Details