Gene Transcripts
- Gene Symbol
- DBET
- Entrez Gene ID
- 100419743
- Gene Name
- D4Z4 binding element transcript (non-protein coding)
- Gene Aliases
- DBE-T, DUX4L30
- Chromosome Location
- Chr. 4: 190064502 - 190067864 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_121644.1 |
|
1 |
1313 |
|
GenBank mRNA |
JQ639078.1 |
- |
2 |
1311 |
|
- Gene Symbol
- DUX4
- Entrez Gene ID
- 100288687
- Gene Name
- double homeobox 4
- Gene Aliases
- DUX4L
- Chromosome Location
- Chr. 4: 190173701 - 190175845 on Build GRCh38
- UniGene ID
- Hs.728749
- Species
- Homo sapiens
View Full Product Details