Gene Transcripts

Gene Symbol
SPDYE5
Entrez Gene ID
442590
Gene Name
speedy/RINGO cell cycle regulator family member E5
Gene Aliases
Chromosome Location
Chr. 7: 75493627 - 75504304 on Build GRCh38
UniGene ID
Hs.632298
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306141.1 NP_001293070.1 6 1508

View Full Product Details