Gene Transcripts
- Gene Symbol
- TMA7
- Entrez Gene ID
- 51372
- Gene Name
- translation machinery associated 7 homolog
- Gene Aliases
- CCDC72, HSPC016
- Chromosome Location
- Chr. 3: 48440276 - 48444129 on Build GRCh38
- UniGene ID
- Hs.356440
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF161448.1 |
- |
3 |
456 |
|
AK126594.1 |
- |
4 |
2367 |
|
AL712164.1 |
- |
4 |
484 |
|
View Full Product Details