Gene Transcripts

Gene Symbol
GAGE12J
Entrez Gene ID
729396
Gene Name
G antigen 12J
Gene Aliases
GAGE11
Chromosome Location
Chr. X: 49322030 - 49329387 on Build GRCh38
UniGene ID
Hs.645554
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098406.2 NP_001091876.2 2 164

View Full Product Details