Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_027307.1 | 2 | 529 | ||
NR_027308.1 | 2 | 529 | |||
GenBank mRNA | AK301086.1 | - | 2 | 96 | |
AK316328.1 | - | 2 | 96 | ||
X63380.1 | - | 2 | 66 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145783.1 | NP_001139255.1 | 2 | 529 | |
NM_001145784.1 | NP_001139256.1 | 2 | 529 | ||
GenBank mRNA | BC000489.2 | - | 2 | 78 | |
BC004449.1 | - | 2 | 78 | ||
BC010178.2 | - | 2 | 87 | ||
BC010931.1 | - | 2 | 94 | ||
BC157837.1 | - | 2 | 360 | ||
BC157868.1 | - | 2 | 142 | 9057087 | |
BC171829.1 | - | 2 | 360 | ||
BC171869.1 | - | 2 | 142 |