Gene Transcripts
- Gene Symbol
- SLC17A2
- Entrez Gene ID
- 10246
- Gene Name
- solute carrier family 17 member 2
- Gene Aliases
- NPT3
- Chromosome Location
- Chr. 6: 25912754 - 25930727 on Build GRCh38
- UniGene ID
- Hs.591802
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U90544.1 |
- |
8 |
1292 |
|
View Full Product Details