Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133664.3 | NP_598425.2 | 8, 9 | 1883 | |
GenBank mRNA | AK083526.1 | - | 8, 9 | 1884 | |
BC016257.1 | - | 8, 9 | 1554 | 4912735 | |
BC025580.1 | - | 8, 9 | 1787 | 5325039 | |
BC031131.1 | - | 8, 9 | 1730 | 4954891 |