Gene Transcripts

Gene Symbol
Snrpc
Entrez Gene ID
20630
Gene Name
U1 small nuclear ribonucleoprotein C
Gene Aliases
Snrp1c, U1-C, U1C
Chromosome Location
Chr. 17: 27840087 - 27851968 on Build GRCm38
UniGene ID
Mm.103949
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011432.2 NP_035562.1 6 667
GenBank mRNA AK137213.1 - 6 650
AK168973.1 - 6 640
BC008243.1 - 6 632 2655238
BC067386.1 - 6040
BC092266.1 - 6 632 4009300
U70315.1 - 6 636
X96767.1 - 6 623

View Full Product Details