Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081756.1 | NP_001075225.1 | 10 | 1126 | |
NM_172484.3 | NP_766072.2 | 8 | 987 | ||
NM_176957.3 | NP_795931.2 | 10 | 1126 | ||
GenBank mRNA | AK053243.1 | - | 8 | 809 | |
AY957601.1 | - | 6 | 546 | ||
AY957602.1 | - | 6 | 546 | ||
BC066095.1 | - | 8 | 987 | 6856184 | |
BC079551.1 | - | 8 | 987 | 6856184 |