Gene Transcripts

Gene Symbol
Rgsl1
Entrez Gene ID
240816
Gene Name
regulator of G-protein signaling like 1
Gene Aliases
4930415K13Rik, Gm202, Rgsl2
Chromosome Location
Chr. 1: 153777929 - 153847342 on Build GRCm38
UniGene ID
Mm.329340
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243223.1 NP_001230152.1 19, 20 3162
GenBank mRNA AK029630.1 - 10, 11 1269

View Full Product Details