Gene Transcripts

Gene Symbol
Ubqln3
Entrez Gene ID
244178
Gene Name
ubiquilin 3
Gene Aliases
4933400K24Rik
Chromosome Location
Chr. 7: 104140621 - 104143321 on Build GRCm38
UniGene ID
Mm.334092
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198623.2 NP_941025.1 2 1453
GenBank mRNA AK077072.1 - 2 1455
BC100419.1 - 2 1430 30918401
BC150726.1 - 1 1380 9087637

View Full Product Details