Gene Transcripts

Gene Symbol
Olfr91
Entrez Gene ID
258470
Gene Name
olfactory receptor 91
Gene Aliases
MOR256-20, bM573K1.3
Chromosome Location
Chr. 17: 37092934 - 37093872 on Build GRCm38
UniGene ID
Mm.377519
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182714.2 NP_874373.2 1 696
GenBank mRNA BC108941.1 - 1 694
BC108942.1 - 1 691
BC119413.1 - 1 696 40042089
BC119414.1 - 1 696 40042091

View Full Product Details