Gene Transcripts

Gene Symbol
Defb34
Entrez Gene ID
360211
Gene Name
defensin beta 34
Gene Aliases
BD-34
Chromosome Location
Chr. 8: 19123752 - 19126540 on Build GRCm38
UniGene ID
Mm.271557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183035.1 NP_898856.1 2 161
GenBank mRNA AJ578469.1 - 2 161

View Full Product Details