Gene Transcripts
- Gene Symbol
- CNPY3
- Entrez Gene ID
- 10695
- Gene Name
- canopy FGF signaling regulator 3
- Gene Aliases
- CAG4A, ERDA5, PRAT4A, TNRC5
- Chromosome Location
- Chr. 6: 42928002 - 42939287 on Build GRCh38
- UniGene ID
- Hs.414099
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318848.1 |
NP_001305777.1
|
2, 3 |
725 |
|
NM_006586.4 |
NP_006577.2
|
2, 3 |
725 |
|
NR_134885.1 |
|
2, 3 |
822 |
|
GenBank mRNA |
AY358960.1 |
- |
2, 3 |
620 |
|
BC004423.1 |
- |
2, 3 |
671 |
2819660 |
BC008898.2 |
- |
2, 3 |
352 |
2959532 |
BC008961.2 |
- |
2, 3 |
352 |
2959532 |
BQ680146.1 |
- |
2, 3 |
344 |
|
CN302713.1 |
- |
2, 3 |
463 |
|
U80744.1 |
- |
2, 3 |
671 |
|
View Full Product Details