Gene Transcripts

Gene Symbol
LILRA3
Entrez Gene ID
11026
Gene Name
leukocyte immunoglobulin like receptor A3
Gene Aliases
CD85E, HM31, HM43, ILT-6, ILT6, LIR-4, LIR4
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.113277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172654.2 NP_001166125.1 8 1354
NM_006865.4 NP_006856.3 8 1590
GenBank mRNA AF014923.1 - 8 1498
AF014924.1 - 8 1306
AF025527.1 - 8 1591
BC028208.1 - 8 1563 5215423
U91926.1 - 8 1560
U91927.1 - 6 1441

View Full Product Details