Gene Transcripts

Gene Symbol
C14orf1
Entrez Gene ID
11161
Gene Name
chromosome 14 open reading frame 1
Gene Aliases
ERG28, NET51
Chromosome Location
Chr. 14: 75650890 - 75661195 on Build GRCh38
UniGene ID
Hs.15106
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007176.3 NP_009107.1 2 496
GenBank mRNA AF134159.2 - 2 116
AF136971.1 - 2 156
AF161406.1 - 2 156
AK311764.1 - 2 136
AL136658.1 - 2 154
BC002444.2 - 2 163 3347242
BX248022.1 - 2 490
CR533443.1 - 1 44

View Full Product Details