Gene Transcripts

Gene Symbol
KLHL20
Entrez Gene ID
27252
Gene Name
kelch like family member 20
Gene Aliases
KHLHX, KLEIP, KLHLX
Chromosome Location
Chr. 1: 173714912 - 173786826 on Build GRCh38
UniGene ID
Hs.495035
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014458.3 NP_055273.2 4 896
XM_005245093.3 XP_005245150.1 5 1045
XM_011509420.2 XP_011507722.1 4 925
XM_017001052.1 XP_016856541.1 4 1175
XM_017001053.1 XP_016856542.1 4 1175
XM_017001054.1 XP_016856543.1 5 1045
GenBank mRNA AB026190.1 - 4 837
AJ844466.1 - 3 717
AK001430.1 - 4 853
AK300755.1 - 3 293
AK308048.1 - 3 814
BC063418.1 - 4 826 5221643

View Full Product Details