Gene Transcripts

Gene Symbol
MMP26
Entrez Gene ID
56547
Gene Name
matrix metallopeptidase 26
Gene Aliases
Chromosome Location
Chr. 11: 4988194 - 4992429 on Build GRCh38
UniGene ID
Hs.204732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021801.4 NP_068573.2 5 641
XM_011520219.2 XP_011518521.1 6 821
GenBank mRNA AF230354.1 - 5 625
AF248646.1 - 5 629
AF291664.1 - 5 638
AJ251531.1 - 5 635
BC101541.1 - 5 641 8069047
BC101543.1 - 5 641 8069049

View Full Product Details