Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282161.1 | NP_001269090.1 | 5 | 499 | |
NM_003336.3 | NP_003327.2 | 5 | 506 | ||
NM_181762.2 | NP_861427.1 | 4 | 416 | ||
GenBank mRNA | AK223045.1 | - | 5 | 408 | |
AK223064.1 | - | 5 | 405 | ||
AK297696.1 | - | 4 | 318 | ||
AK301675.1 | - | 4 | 417 | ||
AK313092.1 | - | 5 | 410 | ||
BC010175.2 | - | 5 | 399 | 3051041 | |
BC042021.1 | - | 2 | 1283 | ||
BI093289.1 | - | 4 | 206 | ||
DA068506.1 | - | 5 | 403 | ||
DA599154.1 | - | 5 | 506 | ||
M74524.1 | - | 5 | 381 |