Gene Transcripts

Gene Symbol
PAGE2
Entrez Gene ID
203569
Gene Name
PAGE family member 2
Gene Aliases
CT16.4, GAGEC2, GAGEE2, PAGE-2
Chromosome Location
Chr. X: 55089052 - 55092836 on Build GRCh38
UniGene ID
Hs.662489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207339.3 NP_997222.1 5 460
XM_017029353.1 XP_016884842.1 5 489
GenBank mRNA BC054022.1 - 5 454
BX113994.1 - 5 390

Gene Symbol
PAGE2B
Entrez Gene ID
389860
Gene Name
PAGE family member 2B
Gene Aliases
CT16.5, GAGEE3, PAGE-2B
Chromosome Location
Chr. X: 55027927 - 55078903 on Build GRCh38
UniGene ID
Hs.293317
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015038.2 NP_001015038.1 5 467
XM_011530785.2 XP_011529087.1 6 883
XM_011530786.2 XP_011529088.1 5 550
XM_011530787.2 XP_011529089.1 5 530
XM_017029513.1 XP_016885002.1 5 487
GenBank mRNA BM782405.1 - 5 452

View Full Product Details