Gene Transcripts
- Gene Symbol
- SLBP
- Entrez Gene ID
- 7884
- Gene Name
- stem-loop binding protein
- Gene Aliases
- HBP
- Chromosome Location
- Chr. 4: 1692731 - 1712741 on Build GRCh38
- UniGene ID
- Hs.298345
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001306074.1 |
NP_001293003.1
|
5, 6 |
1077 |
|
NM_001306075.1 |
NP_001293004.1
|
5, 6 |
1065 |
|
NM_006527.3 |
NP_006518.1
|
6, 7 |
1182 |
|
GenBank mRNA |
AK091735.1 |
- |
5, 6 |
642 |
|
AK093314.1 |
- |
6, 7 |
655 |
|
AK094257.1 |
- |
6, 7 |
996 |
|
AK300826.1 |
- |
5, 6 |
655 |
|
AK315635.1 |
- |
6, 7 |
753 |
|
BC014908.1 |
- |
6, 7 |
751 |
3908584 |
BC015703.1 |
- |
6, 7 |
751 |
3907984 |
BT007433.1 |
- |
6, 7 |
629 |
|
U75679.1 |
- |
6, 7 |
744 |
|
Z71188.1 |
- |
6, 7 |
733 |
|
View Full Product Details