Gene Transcripts

Gene Symbol
PHLPP2
Entrez Gene ID
23035
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
PHLPPL, PPM3B
Chromosome Location
Chr. 16: 71644924 - 71724701 on Build GRCh38
UniGene ID
Hs.709458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289003.1 NP_001275932.1 18 3520
NM_015020.3 NP_055835.2 19 3374
GenBank mRNA AB023148.2 - 18 3520
BC129927.1 - 18 3001 40124720
BX647823.1 - 19 3375
BX648157.1 - 13 2110

View Full Product Details