Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015024.4 | NP_055839.3 | 27 | 3252 | |
XM_017013244.1 | XP_016868733.1 | 27 | 3506 | ||
XM_017013245.1 | XP_016868734.1 | 27 | 3273 | ||
XM_017013246.1 | XP_016868735.1 | 27 | 3475 | ||
GenBank mRNA | AB018288.1 | - | 24 | 2616 | |
AF064729.1 | - | 27 | 3162 | ||
AK291472.1 | - | 27 | 3167 | ||
AK299848.1 | - | 23 | 2688 | ||
BC014219.2 | - | 4 | 511 | ||
BC030785.1 | - | 27 | 3251 | 5269230 |