Gene Transcripts

Gene Symbol
HIST3H2A
Entrez Gene ID
92815
Gene Name
histone cluster 3, H2a
Gene Aliases
Chromosome Location
Chr. 1: 228457364 - 228457859 on Build GRCh38
UniGene ID
Hs.26331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033445.2 NP_254280.1 1 164
GenBank mRNA AK311930.1 - 1 176
BC001193.1 - 1 164
BC082269.1 - 1 154

View Full Product Details