Gene Transcripts

Gene Symbol
NUDT7
Entrez Gene ID
283927
Gene Name
nudix hydrolase 7
Gene Aliases
Chromosome Location
Chr. 16: 77722488 - 77742260 on Build GRCh38
UniGene ID
Hs.282665
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105663.2 NP_001099133.1 3, 4 435
XM_011523032.2 XP_011521334.1 4, 5 553
GenBank mRNA AA227330.1 - 3, 4 387
BU579534.1 - 3, 4 381
ES309123.1 - 1, 2 170

View Full Product Details