Gene Transcripts

Gene Symbol
PLXND1
Entrez Gene ID
23129
Gene Name
plexin D1
Gene Aliases
PLEXD1
Chromosome Location
Chr. 3: 129555213 - 129606739 on Build GRCh38
UniGene ID
Hs.301685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015103.2 NP_055918.2 33 5489
XM_011512588.1 XP_011510890.1 33 5404
XM_011512589.1 XP_011510891.1 30 5014
XM_011512592.1 XP_011510894.1 21 2572
GenBank mRNA AB014520.2 - 33 5568
AK026388.1 - 9 986
AK299425.1 - 12 1324
AY116661.1 - 33 5569
BC003526.1 - 11 1236
BC011848.2 - 10 1179
BC044228.1 - 1 214
BC150280.1 - 33 5568 9007263

View Full Product Details