Gene Transcripts

Gene Symbol
Aff3
Entrez Gene ID
16764
Gene Name
AF4/FMR2 family, member 3
Gene Aliases
3222402O04Rik, A730046J16, LAF-4, Laf4
Chromosome Location
Chr. 1: 38175991 - 38627837 on Build GRCm38
UniGene ID
Mm.336679
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290814.1 NP_001277743.1 13 2680
GenBank mRNA AK014322.1 - 10 1987

View Full Product Details