Gene Transcripts

Gene Symbol
Mup1
Entrez Gene ID
17840
Gene Name
major urinary protein 1
Gene Aliases
Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
Chromosome Location
Chr. 4: 60498012 - 60501960 on Build GRCm38
UniGene ID
Mm.335875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163010.1 NP_001156482.1 2 259
NM_001163011.1 NP_001156483.1 2 259
NM_031188.2 NP_112465.2 2 259
GenBank mRNA BC012221.1 - 2 205
BC013649.1 - 193 4195401
BC091744.1 - 196 5102271
BC091775.1 - 2 206 5137341
BC092254.1 - 193 5100458
BC093515.1 - 193
BC099597.1 - 196 947916
BC106092.1 - 2 196 5098340
M16355.1 - 2 259
M27609.1 - 61
M28649.1 - 206
X03524.1 - 61
X04115.1 - 17

Gene Symbol
LOC100048884
Entrez Gene ID
100048884
Gene Name
novel member of the major urinary protein (Mup) gene family
Gene Aliases
Mup6
Chromosome Location
Chr. 4: 61670177 - 61674098 on Build GRCm38
UniGene ID
Mm.335875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199333.1 NP_001186262.1 2 220
GenBank mRNA BC055353.1 - 195

Gene Symbol
Mup10
Entrez Gene ID
100039008
Gene Name
major urinary protein 10
Gene Aliases
2610016E04Rik, Mup8
Chromosome Location
Chr. 4: 60578260 - 60582155 on Build GRCm38
UniGene ID
Mm.335875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122647.1 NP_001116119.1 2 215
GenBank mRNA AJ309921.1 - 148
CB948918.1 - 124
EU882231.1 - 2 148

Gene Symbol
Mup11
Entrez Gene ID
100039028
Gene Name
major urinary protein 11
Gene Aliases
Gm12549, OTTMUSG00000007431
Chromosome Location
Chr. 4: 60658466 - 60662395 on Build GRCm38
UniGene ID
Mm.482557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164526.1 NP_001157998.1 246
GenBank mRNA AK013972.1 - 2 223

Gene Symbol
Mup12
Entrez Gene ID
100039054
Gene Name
major urinary protein 12
Gene Aliases
Gm2024
Chromosome Location
Chr. 4: 60737381 - 60741281 on Build GRCm38
UniGene ID
Mm.482236
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199995.1 NP_001186924.1 2 214

Gene Symbol
Mup14
Entrez Gene ID
100039116
Gene Name
major urinary protein 14
Gene Aliases
Gm13514, Mup12, OTTMUSG00000012493
Chromosome Location
Chr. 4: 61300037 - 61303998 on Build GRCm38
UniGene ID
Mm.458441
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199999.1 NP_001186928.1 2 257

Gene Symbol
Mup15
Entrez Gene ID
100039150
Gene Name
major urinary protein 15
Gene Aliases
Gm2068, Mup13
Chromosome Location
Chr. 4: 61435787 - 61439714 on Build GRCm38
UniGene ID
Mm.237772
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001200004.1 NP_001186933.1 2 220

Gene Symbol
Mup16
Entrez Gene ID
100039177
Gene Name
major urinary protein 16
Gene Aliases
Chromosome Location
Chr. 4: 61515592 - 61519503 on Build GRCm38
UniGene ID
Mm.422695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199936.1 NP_001186865.1 2 214

Gene Symbol
Mup19
Entrez Gene ID
100189605
Gene Name
major urinary protein 19
Gene Aliases
100039247, Gm12552, Mup11, Mup14, Mup17, Mup8, OTTMUSG00000007472
Chromosome Location
Chr. 4: 61778324 - 61782236 on Build GRCm38
UniGene ID
Mm.457980
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135127.2 NP_001128599.1 2 214
GenBank mRNA AK013259.1 - 2 214
EU882232.1 - 2 142

Gene Symbol
Mup2
Entrez Gene ID
17841
Gene Name
major urinary protein 2
Gene Aliases
AA589603, Mup-2, Mup4
Chromosome Location
Chr. 4: 60135913 - 60139872 on Build GRCm38
UniGene ID
Mm.335875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045550.2 NP_001039015.1 2 218
NM_008647.4 NP_032673.3 2 218
GenBank mRNA BC012259.1 - 207
BC059097.1 - 193 4159984
CB593503.1 - 216
M16356.1 - 206

Gene Symbol
Mup8
Entrez Gene ID
100041687
Gene Name
major urinary protein 8
Gene Aliases
Gm12809, Mup5, OTTMUSG00000008509
Chromosome Location
Chr. 4: 60218620 - 60222599 on Build GRCm38
UniGene ID
Mm.457982
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134676.1 NP_001128148.1 2 278

Gene Symbol
Mup9
Entrez Gene ID
100038948
Gene Name
major urinary protein 9
Gene Aliases
Gm14076, Mup2, Mup6, OTTMUSG00000015595
Chromosome Location
Chr. 4: 60418046 - 60421961 on Build GRCm38
UniGene ID
Mm.482231
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281979.1 NP_001268908.1 2 214
GenBank mRNA BI219655.1 - 2 195


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BC094452.1 - 194 Chr. - - [-]

View Full Product Details