Gene Transcripts

Gene Symbol
Nkx2-8
Entrez Gene ID
299061
Gene Name
NK2 homeobox 8
Gene Aliases
Nkx2-9
Chromosome Location
Chr. 6: 77506882 - 77508585 on Build Rnor_6.0
UniGene ID
Rn.218683
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106732.1 NP_001100202.1 1 277

View Full Product Details