Gene Transcripts

Gene Symbol
Ms4a12
Entrez Gene ID
293741
Gene Name
membrane-spanning 4-domains, subfamily A, member 12
Gene Aliases
Chromosome Location
Chr. 1: 227404994 - 227417504 on Build Rnor_6.0
UniGene ID
Rn.81309
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006231100.2 XP_006231162.1 7 1808
XM_008760272.1 XP_008758494.1 7 1701

View Full Product Details