Gene Transcripts

Gene Symbol
Prlh
Entrez Gene ID
63850
Gene Name
prolactin releasing hormone
Gene Aliases
Prh, prrp
Chromosome Location
Chr. 9: 98111421 - 98114830 on Build Rnor_6.0
UniGene ID
Rn.91303
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022222.1 NP_071558.1 2 116
XM_006245478.2 XP_006245540.1 3 820
GenBank mRNA AB015418.1 - 2 116
AF521930.1 - 2 180

View Full Product Details