Gene Transcripts

Gene Symbol
Mln
Entrez Gene ID
100126231
Gene Name
motilin
Gene Aliases
Chromosome Location
Chr. 20: 6567092 - 6571791 on Build Rnor_6.0
UniGene ID
Rn.218365
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110056.1 NP_001103526.1 1 91
GenBank mRNA AB092487.1 - 1 99
AB111861.1 - 1 91

View Full Product Details