Gene Transcripts

Gene Symbol
Fsd1
Entrez Gene ID
301506
Gene Name
fibronectin type III and SPRY domain containing 1
Gene Aliases
Chromosome Location
Chr. 9: 11076670 - 11080190 on Build Rnor_6.0
UniGene ID
Rn.45600
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106918.1 NP_001100388.1 2, 3 280
GenBank mRNA BC160924.1 - 120

View Full Product Details