Gene Transcripts

Gene Symbol
Lrrc24
Entrez Gene ID
362945
Gene Name
leucine rich repeat containing 24
Gene Aliases
RGD1308720
Chromosome Location
Chr. 7: 117779733 - 117786872 on Build Rnor_6.0
UniGene ID
Rn.161989
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135896.1 NP_001129368.1 1 82
GenBank mRNA BC127521.1 - 1 82

View Full Product Details