Gene Transcripts

Gene Symbol
NUDCD3
Entrez Gene ID
23386
Gene Name
NudC domain containing 3
Gene Aliases
NudCL
Chromosome Location
Chr. 7: 44382366 - 44490875 on Build GRCh38
UniGene ID
Hs.488171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015332.3 NP_056147.2 5 1114
XM_011515247.2 XP_011513549.1 5 1203
XM_017011909.1 XP_016867398.1 6 1140
XM_017011910.1 XP_016867399.1 6 998
GenBank mRNA AB028991.1 - 5 908
AK024226.1 - 4 622
AK127163.1 - 515
AK307613.1 - 3 421
BC003691.2 - 5 975 3453305
BC011673.2 - 4 734
BC017657.1 - 6 1063
BC035014.2 - 5 1012 4812229
EU350932.1 - 5 932

View Full Product Details