Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
Chromosome Location
Chr. 9: 71683366 - 71769532 on Build GRCh38
UniGene ID
Hs.494146
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135820.1 NP_001129292.1 17, 18 3554
NM_013390.2 NP_037522.1 18, 19 3743
XM_005251869.4 XP_005251926.1 18, 19 3376
GenBank mRNA AB037833.2 - 18, 19 3336
AF137030.1 - 18, 19 3351
AK075370.1 - 11, 12 1471
AK303765.1 - 1, 2 242
BC140773.1 - 17, 18 3056 9021641
BC144510.1 - 18, 19 3211
BC146780.1 - 18, 19 3336 9007233
BC171782.1 - 17, 18 3057

View Full Product Details