Gene Transcripts

Gene Symbol
CASP10
Entrez Gene ID
843
Gene Name
caspase 10
Gene Aliases
ALPS2, FLICE2, MCH4
Chromosome Location
Chr. 2: 201182881 - 201229406 on Build GRCh38
UniGene ID
Hs.5353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206524.1 NP_001193453.1 7 1178
NM_001206542.1 NP_001193471.1 7 1250
NM_001230.4 NP_001221.2 7 1250
NM_032974.4 NP_116756.2 9 1379
NM_032976.3 NP_116758.1 8 1287
NM_032977.3 NP_116759.2 9 1379
XM_005246907.2 XP_005246964.1 9 1162
XM_006712796.3 XP_006712859.1 8 1297
XM_011511990.2 XP_011510292.1 8 1070
GenBank mRNA AF111344.1 - 7 869
AF111345.1 - 8 961
AJ487678.1 - 7 884
AJ487679.1 - 7 953
BC042844.1 - 9 1115 5176698
KU177959.1 - 8 961
KU177960.1 - 6 832
U60519.1 - 7 979
U86214.1 - 9 1155

View Full Product Details