Gene Transcripts
- Gene Symbol
- TIPARP
- Entrez Gene ID
- 25976
- Gene Name
- TCDD inducible poly(ADP-ribose) polymerase
- Gene Aliases
- ARTD14, PARP7, pART14
- Chromosome Location
- Chr. 3: 156674416 - 156706770 on Build GRCh38
- UniGene ID
- Hs.744050
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001184717.1 |
NP_001171646.1
|
3, 4 |
1597 |
|
NM_001184718.1 |
NP_001171647.1
|
3, 4 |
1162 |
|
NM_015508.4 |
NP_056323.2
|
3, 4 |
1512 |
|
GenBank mRNA |
AK097785.1 |
- |
3, 4 |
1341 |
|
AK291261.1 |
- |
3, 4 |
1347 |
|
AL080156.1 |
- |
2, 3 |
198 |
|
BC034397.1 |
- |
3, 4 |
1162 |
|
BC050350.1 |
- |
3, 4 |
1319 |
5264807 |
BX537965.1 |
- |
3, 4 |
1339 |
|
CR749647.1 |
- |
3, 4 |
1598 |
|
View Full Product Details