Gene Transcripts
- Gene Symbol
- DKK3
- Entrez Gene ID
- 27122
- Gene Name
- dickkopf WNT signaling pathway inhibitor 3
- Gene Aliases
- REIC, RIG
- Chromosome Location
- Chr. 11: 11962996 - 12009370 on Build GRCh38
- UniGene ID
- Hs.292156
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001018057.1 |
NP_001018067.1
|
2, 3 |
401 |
|
NM_013253.4 |
NP_037385.2
|
3, 4 |
569 |
|
NM_015881.5 |
NP_056965.3
|
3, 4 |
583 |
|
XM_006718178.2 |
XP_006718241.1
|
3, 4 |
396 |
|
XM_017017553.1 |
XP_016873042.1
|
3, 4 |
530 |
|
XM_017017554.1 |
XP_016873043.1
|
3, 4 |
548 |
|
XM_017017555.1 |
XP_016873044.1
|
3, 4 |
396 |
|
GenBank mRNA |
AB033421.1 |
- |
2, 3 |
362 |
|
AB034203.1 |
- |
3, 4 |
541 |
|
AB057591.1 |
- |
3, 4 |
569 |
|
AF177396.1 |
- |
2, 3 |
381 |
|
AK090952.1 |
- |
|
488 |
|
AK092979.1 |
- |
3, 4 |
360 |
|
AK225156.1 |
- |
2, 3 |
353 |
|
AK289897.1 |
- |
3, 4 |
410 |
|
AK295539.1 |
- |
3, 4 |
504 |
|
AL535720.3 |
- |
2, 3 |
400 |
|
AY358378.1 |
- |
3, 4 |
496 |
|
AY587550.1 |
- |
2, 3 |
344 |
|
BC007660.2 |
- |
3, 4 |
582 |
3347453 |
View Full Product Details