Gene Transcripts

Gene Symbol
KLHL20
Entrez Gene ID
27252
Gene Name
kelch like family member 20
Gene Aliases
KHLHX, KLEIP, KLHLX
Chromosome Location
Chr. 1: 173714912 - 173786826 on Build GRCh38
UniGene ID
Hs.495035
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014458.3 NP_055273.2 9 1517
XM_005245093.3 XP_005245150.1 10 1666
XM_011509420.2 XP_011507722.1 8 1402
XM_017001052.1 XP_016856541.1 9 1796
XM_017001053.1 XP_016856542.1 8 1652
XM_017001054.1 XP_016856543.1 9 1522
GenBank mRNA AB026190.1 - 9 1458
AJ844466.1 - 8 1338
AK001430.1 - 9 1474
AK300755.1 - 8 914
AK308048.1 - 8 1435
BC063418.1 - 9 1447 5221643

View Full Product Details