Gene Transcripts

Gene Symbol
PURG
Entrez Gene ID
29942
Gene Name
purine rich element binding protein G
Gene Aliases
PURG-A, PURG-B
Chromosome Location
Chr. 8: 30995802 - 31033715 on Build GRCh38
UniGene ID
Hs.373778
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015508.2 NP_001015508.1 1 1776
NM_001323311.1 NP_001310240.1 2 966
NM_001323312.1 NP_001310241.1 2 966
NM_013357.2 NP_037489.1 1 862
GenBank mRNA AF195513.2 - 1 1776
AY077841.1 - 1 1776
BC106708.2 - 1 862 40005076

View Full Product Details