Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016024.2 | NP_057108.2 | 5 | 386 | |
GenBank mRNA | AF078865.1 | - | 4 | 322 | |
AF151837.1 | - | 4 | 322 | ||
AK292855.1 | - | 5 | 368 | ||
AL050405.1 | - | 5 | 361 | ||
BC003676.1 | - | 5 | 386 | ||
BC033750.1 | - | 5 | 354 | 5210341 | |
BC040433.1 | - | 5 | 337 | ||
BC105582.1 | - | 5 | 373 |