Gene Transcripts

Gene Symbol
IL17D
Entrez Gene ID
53342
Gene Name
interleukin 17D
Gene Aliases
IL-17D
Chromosome Location
Chr. 13: 20702775 - 20723100 on Build GRCh38
UniGene ID
Hs.655142
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138284.1 NP_612141.1 3 1104
XM_005266421.3 XP_005266478.1 3 1548
XM_005266422.2 XP_005266479.1 2 1265
GenBank mRNA AF458062.1 - 3 1054
AK123964.1 - 3 1187
AY078238.1 - 3 1104
AY359113.1 - 2 996
BC036243.1 - 3 1107 5301608

View Full Product Details