Gene Transcripts

Gene Symbol
MIEF1
Entrez Gene ID
54471
Gene Name
mitochondrial elongation factor 1
Gene Aliases
HSU79252, MID51, SMCR7L, dJ1104E15.3
Chromosome Location
Chr. 22: 39500100 - 39518134 on Build GRCh38
UniGene ID
Hs.728085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304564.1 NP_001291493.1 6 1408
NM_019008.5 NP_061881.2 6 1458
NR_130789.1 6 1372
NR_130790.1 7 1522
XM_011530236.1 XP_011528538.1 8 1526
XM_011530237.1 XP_011528539.1 7 1646
XM_011530238.1 XP_011528540.1 7 1730
XM_017028838.1 XP_016884327.1 7 1624
GenBank mRNA AK000239.1 - 3 629
AK290954.1 - 7 1355
AL136768.1 - 6 1408
AL365515.1 - 6 1335
AL834205.1 - 6 1300
BC002587.2 - 2 600
BC008327.1 - 6 1312 3502711

View Full Product Details