Gene Transcripts

Gene Symbol
MS4A12
Entrez Gene ID
54860
Gene Name
membrane spanning 4-domains A12
Gene Aliases
Ms4a10
Chromosome Location
Chr. 11: 60492743 - 60507430 on Build GRCh38
UniGene ID
Hs.272789
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164470.1 NP_001157942.1 4 409
NM_017716.2 NP_060186.2 5 547
XM_011545117.2 XP_011543419.1 6 695
GenBank mRNA AK000224.1 - 5 547
BC029793.1 - 5 535 5185424
BP262076.1 - 5 549
BP262976.1 - 4 409

View Full Product Details