Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017752.2 | NP_060222.2 | 10 | 1696 | |
NM_198881.1 | NP_942582.1 | 10 | 1696 | ||
XM_006724669.1 | XP_006724732.1 | 9 | 1546 | ||
XM_006724670.3 | XP_006724733.1 | 10 | 1510 | ||
GenBank mRNA | AB449890.1 | - | 10 | 1531 | |
AB449891.1 | - | 10 | 1531 | ||
AK091248.1 | - | 1 | 13 | ||
AK123957.1 | - | 10 | 1696 | ||
BC122564.1 | - | 10 | 1695 | 40026331 |