Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018318.3 | NP_060788.3 | 4 | 447 | |
XM_005253413.1 | XP_005253470.1 | 5 | 660 | ||
XM_005253414.1 | XP_005253471.1 | 5 | 520 | ||
XM_005253415.1 | XP_005253472.1 | 4 | 589 | ||
XM_006719103.3 | XP_006719166.1 | 4 | 611 | ||
XM_006719104.2 | XP_006719167.1 | 8 | 842 | ||
XM_006719105.1 | XP_006719168.1 | 6 | 695 | ||
XM_011520729.2 | XP_011519031.1 | 6 | 706 | ||
XM_011520731.2 | XP_011519033.1 | 4 | 704 | ||
XM_011520733.2 | XP_011519035.1 | 4 | 1031 | ||
XM_011520734.1 | XP_011519036.1 | 3 | 508 | ||
XM_011520735.1 | XP_011519037.1 | 3 | 463 | ||
XM_017019569.1 | XP_016875058.1 | 7 | 735 | ||
XM_017019570.1 | XP_016875059.1 | 5 | 629 | ||
XM_017019571.1 | XP_016875060.1 | 6 | 666 | ||
XM_017019572.1 | XP_016875061.1 | 7 | 629 | ||
XM_017019573.1 | XP_016875062.1 | 4 | 732 | ||
XM_017019574.1 | XP_016875063.1 | 6 | 626 | ||
XM_017019575.1 | XP_016875064.1 | 8 | 826 | ||
XM_017019576.1 | XP_016875065.1 | 8 | 842 | ||
XM_017019577.1 | XP_016875066.1 | 5 | 660 | ||
GenBank mRNA | AK093152.1 | - | 7 | 714 | |
AK309181.1 | - | 5 | 520 | ||
AY289196.1 | - | 4 | 447 | ||
BC018679.1 | - | 4 | 865 | ||
BC028682.2 | - | 4 | 523 | 4827882 | |
CR749586.1 | - | 1 | 110 |