Gene Transcripts

Gene Symbol
SLC29A3
Entrez Gene ID
55315
Gene Name
solute carrier family 29 member 3
Gene Aliases
ENT3, HCLAP, HJCD, PHID
Chromosome Location
Chr. 10: 71319253 - 71363390 on Build GRCh38
UniGene ID
Hs.438419
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174098.1 NP_001167569.1 6 1054
NM_018344.5 NP_060814.4 6 1048
NR_033413.1 5 965
NR_033414.1 4 738
XM_006717910.3 XP_006717973.1 6 1440
XM_017016377.1 XP_016871866.1 5 1262
XM_017016378.1 XP_016871867.1 4 1038
GenBank mRNA AF326987.1 - 5 996
AK002022.1 - 5 799
AK304503.1 - 5 935
AK314497.1 - 6 1016
AK316152.1 - 5 950
AX775915.1 - 6 1048
AX775917.1 - 6 1048
AX775919.1 - 6 1048
AX775921.1 - 6 1048
AY288928.1 - 6 1008
AY358686.1 - 6 1026
BC000223.1 - 3 423
BC041575.1 - 5 936
BC120996.1 - 6 1026 40116513
BC120997.1 - 6 1026 40116514

View Full Product Details