Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001276320.1 | NP_001263249.1 | 4 | 809 | |
NM_018566.3 | NP_061036.3 | 2 | 783 | ||
GenBank mRNA | AK128014.1 | - | 4 | 809 | |
AL833081.2 | - | 2 | 715 | ||
BC137166.1 | - | 2 | 775 | 9021163 | |
BC137167.1 | - | 2 | 775 | 9021164 |